Learn More
Fisherbrand™ Foil Roll
Product Code. 11977994 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Width (Metric):
102 mm
153 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
11977994 153 mm Each
11967994 102 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11977994
  • ONLINE EXCLUSIVE PRICE
    251.00 NOK
    221.46 NOK / Pack of 1
    Save 29.54 NOK
    11% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Cut foil to any size length. Foil Roll can be used with the Mini Bin Foil Dispenser or on its own.
  • Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and Petri dishes.
  • Ideal for autocalving, freezing, incubating, protecting labware, lining, sealing, and general storage.
  • 152.4 m of foil available in two widths: 102 mm or 153 mm
  • Disposable
  • Designed to fit the Mini Bin Foil Dispenser.
Specifications
Applications Autoclaving, freezing, incubating, protecting labware, lining, sealing, and general storage
Length (English) 500 ft.
Color Metallic
Product Type Aluminum Foil
Width (English) 6 in.
Width (Metric) 153 mm
Length (Metric) 152.4 m
Material Aluminum
Thickness 15 μm
For Use With (Equipment) Mini Bin Foil Dispenser