Learn More
R&L Enterprises™ Aluminum Alloy Rods
Product Code. 12388139 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
Unit Size:
Pack of 2
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12388139 600 mm Pack of 5
12378139 500 mm Pack of 5
12398139 750 mm Pack of 2
12308149 1000 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12388139
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Aluminum Alloy Rods are manufactured from aluminum alloy.
  • Threaded
  • Thread diameter 10mm
  • Length of rod does not include the thread
Specifications
Material Aluminum Alloy
Length (Metric) 600 mm
Product Type Aluminum Rods
Thread Size 10 mm
Quantity 5 Pk.