missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11032-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
alpha Actin | |
Polyclonal | |
Unconjugated | |
ACTA1 | |
ACTC, ACTC1, ACTC1 cardiac muscle 1, Actin, alpha cardiac muscle 1, actin, alpha, cardiac muscle 1, Alpha actin, Alpha cardiac actin, CMD1R, CMH11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11459, 11464, 29275, 29437, 58, 70 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P68032, P68033, P68035, P68133, P68134, P68136 | |
ACTA1, ACTC1 | |
alpha Actin Fusion Protein Ag1471 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |