missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | alpha Actin |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887823
|
Proteintech
11032-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877823
|
Proteintech
11032-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
alpha Actin | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P68032, P68033, P68035, P68133, P68134, P68136 | |
11459, 11464, 29275, 29437, 58, 70 | |
alpha Actin Fusion Protein Ag1471 | |
Primary | |
-20°C | |
ACTA1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACTC, ACTC1, ACTC1 cardiac muscle 1, Actin, alpha cardiac muscle 1, actin, alpha, cardiac muscle 1, Alpha actin, Alpha cardiac actin, CMD1R, CMH11 | |
ACTA1, ACTC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |