Learn More
Allyl ethyl ether, 98%, Thermo Scientific™
Product Code. 11465936
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11465936 10 g 10g
11475936 50 g 50g
2 options
Product Code. 11465936
Brand: Thermo Scientific Alfa Aesar B22276.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Allyl ethyl ether is used as an intermediate to manufacture other chemicals.
Solubility
Soluble in organic solvents, miscible with ethanol, diethyl ether. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 557-31-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009255
InChI Key OJPSFJLSZZTSDF-UHFFFAOYSA-N
Synonym allyl ethyl ether, 1-propene, 3-ethoxy, allylethylether, ethyl allyl ether, unii-sm8t658re3, ether, allyl ethyl, 3-ethoxy-1-propene, ethyl 2-propenyl ether, 3-ethoxypropene, acmc-1atoi
PubChem CID 11191
IUPAC Name 3-ethoxyprop-1-ene
SMILES CCOCC=C
Specifications
Boiling Point 64°C to 65°C
Flash Point −20°C (−4°F)
Linear Formula CH2=CHCH2OCH2CH3
Quantity 10 g
UN Number UN2335
Beilstein 1361480
Merck Index 14,293
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material Allyl ethyl ether