accessibility menu, dialog, popup

UserName

CAS RN 557-31-3

H3COCH2

CAS RN 557-31-3

IUPAC Name: 3-ethoxyprop-1-ene
Synonyms: allyl ethyl ether
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: OJPSFJLSZZTSDF-UHFFFAOYSA-N
SMILES: CCOCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results