missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline phosphatase from E. coli, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
100 U
200 U
Unit Size:
100 units
1ku
200 units
Description
- It is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection.
- It may be used for protein labeling when high sensitivity is required.
- Alkaline phosphatase is a hydrolase enzyme responsible for removing phosphate groups from many types of molecules, including nucleotides, proteins, and alkaloids.
- It is most effective in an alkaline environment. It is sometimes used synonymously as basic phosphatase.
Specifications
Specifications
| Quantity | 100 U |
| Physical Form | Suspension |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction