accessibility menu, dialog, popup

UserName

CAS RN 9001-78-9

CH3ONONHONONHNNH2H2NOHOH2N

CAS RN 9001-78-9

IUPAC Name: 2-{[1-(2-{[1-(2-aminopropanoyl)pyrrolidin-2-yl]formamido}acetyl)pyrrolidin-2-yl]formamido}-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms: alkaline phosphatase
Molecular Weight (g/mol): 496.57
Molecular Formula: C21H36N8O6
InChi Key: ITZMJCSORYKOSI-UHFFFAOYNA-N
SMILES: CC(N)C(=O)N1CCCC1C(=O)NCC(=O)N1CCCC1C(=O)NC(CCCN=C(N)N)C(O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results