Learn More
Alkaline phosphatase from E. coli, MP Biomedicals™
Product Code. 11481105 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
100 U
200 U
Unit Size:
100 units
1ku
200 units
Product Code. 11481105
Brand: MP Biomedicals 0219981782
Alkaline phosphatase from E. coli ≥30 u/mg protein , Activity- ∽1000 u/mL, Suspension
  • It is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection.
  • It may be used for protein labeling when high sensitivity is required.
  • Alkaline phosphatase is a hydrolase enzyme responsible for removing phosphate groups from many types of molecules, including nucleotides, proteins, and alkaloids.
  • It is most effective in an alkaline environment. It is sometimes used synonymously as basic phosphatase.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9001-78-9
Molecular Formula C21H36N8O6
Molecular Weight (g/mol) 496.57
MDL Number MFCD00131849
InChI Key ITZMJCSORYKOSI-UHFFFAOYNA-N
Synonym Orthophosphoric Monoester Phosphohydrolase
Specifications
Quantity 200 U
Physical Form Suspension
compliance-icons
Product Identifier
  • ALKALINE PHOSPHATASE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium Sulphate