Learn More
Invitrogen™ ALCAM/CD166 Human ELISA Kit
Sandwich ELISA Kit
5350.00 NOK - 26540.00 NOK
Specifications
Accession Number | Q13740 |
---|---|
Assay Sensitivity | 15 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454893
|
Invitrogen™
EHALCAM |
96 Tests |
5350.00 NOK
1 set |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17105349
|
Invitrogen™
EHALCAMX5 |
5 x 96 Tests |
26540.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The Human ALCAM/CD166 (Hu ALCAM) ELISA quantitates Hu ALCAM in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ALCAM. Principle of the method The Human ALCAM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CD166 is an activated leukocyte cell adhesion molecule (ALCAM) which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. CD166 binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. CD166 is involved in neurite extension by neurons via heterophilic and homophilic interactions, and may play a role in the binding of T- and B-cells to activated leukocytes, as well as in interactions between cells of the nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been found for CD166.Specifications
Q13740 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
CD166,MEMD | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
15 pg/mL | |
Biotin | |
CD Protein | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP- 1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
214 | |
<12% | |
HRP | |
Extracellular Matrix & Cell Adhesion | |
CD166, MEMD | |
1 hr. 20 min. |