missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AKT1 Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
This product is intended for use as a positive control in Western blot. You will receive the lysate (100 ug), and an empty vector negative control (100 ug).
The AKT1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | This AKT1 lysate is made in HEK293T cells, using plasmid ID RC201850 and based on accession number NM_001014431. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens v-akt murine thymoma viral oncogene homolog 1 (AKT1), transcript variant 3, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction