Learn More
Uvex™ Airwing B-S-WR Safety Helmet
A lightweight helmet with an attractive design.
Brand:  Uvex™ 9762431
Additional Details : Weight : 11.00000kg
 View more versions of this product
Product Code. 17504106
  • 4985.00 NOK / Pack of 25
Estimated Shipment: 11-06-2024
Description
  • Lightweight safety helmet with short brim and three air vents, one ventilation zone on the forehead and two at the back
  • 6-point textile suspension harness
  • Side slots for attaching ear defenders or visors
  • Rotating size adjusters
Specifications
Specifications
EN 397
HDPE
Industrial Safety Helmet
Short Brim
Green
9762
52 to 61 cm
Ventilated