Learn More
Uvex™ Airwing B-S-WR Safety Helmet
A lightweight helmet with an attractive design.
Brand: Uvex™ 9762131
Additional Details : Weight : 11.00000kg
Description
- Lightweight safety helmet with short brim and three air vents, one ventilation zone on the forehead and two at the back
- 6-point textile suspension harness
- Side slots for attaching ear defenders or visors
- Rotating size adjusters
Specifications
EN 397 | |
HDPE | |
Industrial Safety Helmet | |
Short Brim |
Yellow | |
9762 | |
52 to 61 cm | |
Ventilated |