Learn More
Syngene™ AirClean™ 12 Month Tier 2 Extended Warranty for Standard/Tall Polycarbonate UV Units
Catalog No. 17775914 Alle ansehen Syngene Produkte
Ansicht ändern
Click to view available options
For Use With (Equipment):
Standard/Tall Polycarbonate UV Units (32 in. W)
Standard/Tall Polycarbonate UV Units (48 in. W)
Packungsgröße:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Packungsgröße
17775914 Standard/Tall Polycarbonate UV Units (32 in. W) Each
17705924 Standard/Tall Polycarbonate UV Units (48 in. W) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 17775914 Lieferant Syngene™ Lieferanten-Nr. SYNACAWT32UV
  • ONLINE EXCLUSIVE PRICE
    25170.00 NOK
    19965.06 NOK / Pack of 1
    Save 5204.94 NOK
    20% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The AirClean™ 12 Month Tier 2 Extended Warranty for Standard/Tall Polycarbonate UV Units warranty extension includes filter replacement and confirmation of operation, smart maintenance reminder program, and the disposal of used filters per manufacturers guidelines.
  • AirClean™ 12 Month Extended Warranty for Wide and Tall Standard Polycarbonate Units.
  • Warranty extension includes filter replacement and confirmation of operation, smart maintenance reminder program.
  • Warranty also includes the disposal of used filters per manufacturers guidelines.
Spezifikation
Duration 12 Months
For Use With (Equipment) Standard/Tall Polycarbonate UV Units (32 in. W)