Learn More
STEMCELL Technologies AggreWell™800 24-well Plate Starter Kit
Description
- AggreWell™ plates bring an easy, standardized approach to the generation of cell aggregates, including embryoid bodies (EBs) and spheroids
- EBs and spheroids generated using AggreWell™ plates are consistent in size and shape, and are uniform within and between experiments
- New and improved second-generation AggreWell™ plates are compatible with a variety of cell types, including ES and iPS cells, cancer cells and more
- Enhanced optical characteristics provide crystal clear imaging
- Note: AggreWell™ Rinsing Solution is required for optimal EB and spheroid formation
Specifications
Specifications
| No. of Wells | 24 |
| For Use With (Application) | Differentiation; Spheroid Culture; Toxicity Assay |
| Quantity | Starter Kit |