Learn More
STEMCELL Technologies AggreWell™400 6-well Plate Starter Kit
Product Code. 16448270
Encompass_Preferred
Change view
Click to view available options
:
Starter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16448270 Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16448270 Supplier STEMCELL Technologies Supplier No. 34460
AggreWell™400 6-well Plate Starter Kit contains 2 AggreWell™400 6-well plates and a bottle of AggreWell™ Rinsing Solution.
  • AggreWell™ plates bring an easy, standardized approach to the generation of cell aggregates, including embryoid bodies (EBs) and spheroids
  • EBs and spheroids generated using AggreWell™ plates are consistent in size and shape, and are uniform within and between experiments
  • New and improved second-generation AggreWell™ plates are compatible with a variety of cell types, including ES and iPS cells, cancer cells and more
  • Enhanced optical characteristics provide crystal clear imaging
  • Note: AggreWell™ Rinsing Solution is required for optimal EB and spheroid formation
Specifications
For Use With (Application) Differentiation; Spheroid Culture; Toxicity Assay
Quantity Starter Kit