missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-diphosphate Monosodium Salt, 97.7%, MP Biomedicals™

Product Code. p-4405466 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 11444592

Brand: MP Biomedicals 02190149.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20398-34-9
Molecular Formula C10H15N5O10P2
Molecular Weight (g/mol) 427.20
MDL Number MFCD00066473,MFCD00065470,MFCD00066635
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
Synonym adp monosodium salt, adenosine 5'-diphosphate, sodium salt, adenosine 5'-diphosphate sodium salt
PubChem CID 87704518
IUPAC Name [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 0.977
Quantity 500 mg
Formula Weight 449.19
Physical Form Powder
Chemical Name or Material Adenosine-5'-diphosphate Monosodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.