Learn More
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Icp110
Shop All Memmert ProductsDescription
- Reinforced stainless steel grid
- Max. loading 60 kg
- From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications
Specifications
| Capacity (Metric) Shelf | 60 kg |
| For Use With (Equipment) | Models 110, 160 Incubator |
| Material | Stainless Steel |
| Type | Reinforced Grid |
| Model | 110 |