Learn More
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Icp110
Product Code. 16356997 Shop All Memmert Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
16356997 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16356997 Supplier Memmert™ Supplier No. ICP110E29767
Only null left
Add to basket
Add to basket
  • Reinforced stainless steel grid
  • Max. loading 60 kg
  • From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications
Capacity (Metric) Shelf 60 kg
For Use With (Equipment) Models 110, 160 Incubator
Material Stainless Steel
Type Reinforced Grid
Model 110