Learn More
Memmert™ Addition of Units for SF750plus
Product Code. 17588375 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF750plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17588375 SF750plus Sterilizer Each
1 options
Product Code. 17588375
Brand: Memmert™ SF750plusFDAQ2
Only null left
Add to basket
Add to basket
Integration of one additional unit (up to max. 31 units) into an already existent FDA-software licence (only for units with TwinDISPLAY).
  • Up to max. 31 units
  • Only for units with TwinDISPLAY
Specifications
For Use With (Equipment) SF750plus Sterilizer