Description
- Adapter for Rotor A-4-38
- For centrifugation of blood collection tubes or conical tubes
Specifications
Specifications
| Type | Adapter |
| Tube Type | Monovette |
| Capacity | 9 to 15 mL |
| For Use With (Equipment) | Round bucket in Rotor A-4-38 |
| Bore Size (Metric) | 17.5 mm |
| No. of Places | 16 |
| No. per Pack | 2 Pack |