Learn More
Eppendorf™ Adapters for Rectangular Buckets in A-4-81 Rotor
Shop All Eppendorf Products
Click to view available options
Capacity:
15 mL
180 to 250 mL
2.6 to 7 mL
3 to 15 mL
Unit Size:
Pack of 2
Description
- Rotor adapters
- For rectangluar buckets
- Available in a wide range of sizes to suit various applications
Specifications
Specifications
Type | Adapter |
Tube Type | Round Bottom |
Capacity | 2.6 to 7 mL |
For Use With (Equipment) | 500mL Rectangular Bucket for Rotor A-4-81 |
Bore Size (Metric) | 13 mm |
No. of Places | 18 |
No. per Pack | 2 Pack |
Product Content Correction
Product Title
Eppendorf™ Adapters for Rectangular Buckets in A-4-81 Rotor >