Learn More
Eppendorf™ Adapters For 100mL Rectangular Bucket
Shop All Eppendorf Products
Click to view available options
Capacity:
1.2 to 5 mL
1.5 to 2 mL
15 mL
18 to 120 mL
18 to 30 mL
2.6 to 7 mL
3 to 15 mL
30 to 50 mL
50 mL
50 to 75 mL
7 to 18 mL
Unit Size:
Pack of 2
Description
- Adapters for rectangular buckets
- For use with Rotor A-4-44
- Available in a variety of sizes for a wide range of applications
Specifications
Specifications
| Type | Adapter |
| Capacity | 3 to 15 mL |
| For Use With (Equipment) | 100mL Rectangular Bucket |
| Bore Size (Metric) | 16 mm |
| No. of Places | 28 |
| For Use With (Application) | Standard, Vacutainer, Falcon and micro test tubes |
| No. per Pack | 2 Pack |