Learn More
Eppendorf™ Adapters for 1.5/2.0mL Microcentrifuge Rotors
Product Code. 10227810 Shop All Eppendorf Products
Click to view available options
Capacity:
6 x 0.2 mL PCR Tubes
6 x 0.25 to 0.4 mL Microtainer Tubes
6 x 0.5 to 0.6 mL Microtainer Tubes
6 x 0.5 to 0.6 mL PCR Tubes
For Use With (Application):
0.2mL PCR Tubes
0.4mL Backman Tubes
0.5mL or 0.6mL Microtainer Tubes
Tube Type:
Backman
Eppendorf
PCR
Unit Size:
Pack of 6
Product Code. 10227810
Brand: Eppendorf™ 5425723008
Use these adaptors with a variety of tubes and rotors. Eppendorf™ Adapters work with all 1.5/2.0mL microcentrifuge rotors.
  • Adaptors for rotors
  • Available in packs of 6
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type PCR
Capacity 6 x 0.25 to 0.4 mL Microtainer Tubes
For Use With (Equipment) Rotor F-45-72-8
For Use With (Application) 0.2mL PCR Tubes
No. per Pack 6/Pk.