Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor Product Code.: 15810937
Eppendorf™ Adapter for S-4xUniversal Rotor
Product Code. p-7159251 Shop All Eppendorf Products
7 x 50 mL Skirted Tubes, 7 x 30 mL Oak Ridge Tubes, Skirted Bottom, Oak Ridge, 7/7, Pack of 2
Click to view available options
Capacity:
1 x 1 L Nalgene Bottle
1 x 500 mL Conical Bottle
1 x 750 mL Flat-bottom Bottle
16 x 5 mL Microtubes, 16 x 15 mL Conical Tubes, 1 MTP/DWP
21 x 9 mL Round Bottom Tubes
26 x 7.5 to 12 mL Round Bottom Tubes
30 x 4 to 8 mL Round Bottom Tubes
46 tubes of 12 x 75mm or 5ml, Round Bottom Tubes (FACS)
5 x MTP/DWP/PCR Plates
7 x 50 mL Conical Tubes, 1 x 96/384-well Microplate, 1 x 250 mL Flat-bottom Bottle, 1 x 175 to 225 mL Conical Bottle
7 x 50 mL Skirted Tubes, 7 x 30 mL Oak Ridge Tubes
9 x 50 mL Conical Tubes, 1 Microplate
Tube Type:
Conical
Flat Bottom
Flat/Conical Bottom
Microtube, Conical
PCR
Round Bottom
Round Bottom (FACS)
Skirted Bottom, Oak Ridge
No. of Places:
1
16/16/1
21
26
30
46
5
7/1/1/1
7/7
9/1
Unit Size:
Pack of 2
Product Code. 15810937
Brand: Eppendorf™ 5910764006
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
TRUSTED_SUSTAINABILITY
Specifications
Tube Type Skirted Bottom, Oak Ridge
Capacity 7 x 50 mL Skirted Tubes, 7 x 30 mL Oak Ridge Tubes
For Use With (Equipment) S-4xUniversal Rotor
No. of Places 7/7
No. per Pack 2
Product Content Correction
Product Title
Eppendorf™ Adapter for S-4xUniversal Rotor >