Description
- Suitable for S-4x400 Rotor buckets
- Accommodates selected array of tubes or bottles
Specifications
Specifications
| Tube Type | Round Bottom |
| Capacity | 15 x 4 to 8 mL Round Bottom Tubes |
| Max. Tube Diameter | 13 mm |
| Max. Tube Length | 119 mm |
| For Use With (Equipment) | S-4x400 Bucket |
| Vessels per Adapter | 15 |
| No. of Places | 15 |
| No. per Pack | 2/Pk. |