Learn More
Eppendorf™ Adapter for S-4x1000 Rotor High-Capacity Bucket
Product Code. 15788389 Shop All Eppendorf Products
Click to view available options
Capacity:
52 x 5 mL Round Bottom Tubes (FACS)
Unit Size:
Pack of 2
Product Code. 15788389
Brand: Eppendorf™ 5920724002
Facilitates the use of specific sizes, shapes, and numbers of vessels in S-4x1000 rotor high-capacity buckets.
  • Suitable for S-4x1000 rotor high-capacity buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Tube Type Round Bottom (FACS)
Capacity 52 x 5 mL Round Bottom Tubes (FACS)
Max. Tube Diameter 12 mm
Max. Tube Length 106 mm
For Use With (Equipment) S-4X1000 rotor high-capacity buckets
Vessels per Adapter 52
No. of Places 52
No. per Pack 2/Pk.