Description
- Suitable for S-4xUniversal-Large rotor buckets
- Accommodates selected array of tubes, bottles, or cards
Specifications
Specifications
| Tube Type | Conical, Skirted |
| Capacity | 7 x 50 mL Conical, Skirted Tubes |
| Max. Tube Diameter | 31 mm |
| Max. Tube Length | 141 mm |
| For Use With (Equipment) | S-4xUniversal-Large |
| Vessels per Adapter | 7 |
| No. of Places | 7 |
| No. per Pack | 2/Pk. |