Description
- Suitable for S-4xUniversal-Large rotor buckets
- Accommodates selected array of tubes, bottles, or cards
Specifications
Specifications
| Tube Type | Conical Bottle |
| Capacity | 1 x 250 Conical Bottle |
| Max. Tube Diameter | 62 mm |
| Max. Tube Length | 171 mm |
| For Use With (Equipment) | S-4xUniversal-Large |
| Vessels per Adapter | 1 |
| No. of Places | 1 |
| No. per Pack | 2/Pk. |