Learn More
Eppendorf™ Adapter for Mirco Test Tube
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
1.5 to 2 mL micro test tubes
Unit Size:
Pack of 2
Description
- 2 pieces
- Adapter
Specifications
Specifications
| Type | Adapter |
| For Use With (Equipment) | 1.5 to 2 mL micro test tubes |
| Description | For Rotor S-4xUniversal |
| No. per Pack | 2 |