Learn More
Hettich™ Adapter for Hettich Drum Rotors no. 1161 and 1711
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 1378
Additional Details : Weight : 0.50000kg
Description
- Rotor Carrier for Hettich Rotors
- For Use with MIKRO 220/220 R and ROTINA 380/380 R Centrifuges
- ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards
- MIKRO 220 and 220 R are powerful, compact benchtop centrifuges for processing microlitre tubes of 0.2 to 2.0mL
Specifications
Rotor Adapter | |
21 x 0.5 to 0.8 mL Tubes | |
40 mm | |
21 |
Microtube | |
8 mm | |
Hettich Drum |