Learn More
Hettich™ Adapter for Hettich Drum Rotors no. 1161 and 1711
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
625.00 NOK - 1735.00 NOK
Specifications
Type Rotor Adapter
For Use With (Equipment) Hettich Drum
View More Specs
Products 3
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
10226533
View Documents Promotion Details
Hettich™
1379
32 x 0.2 to 0.4 mL Tubes
1255.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
10530513
View Documents Promotion Details
Hettich™
1377
10 x 1.5 to 2 mL Tubes
625.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
10347923
View Documents Promotion Details
Hettich™
1378
21 x 0.5 to 0.8 mL Tubes
1735.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
Description
  • Rotor Carrier for Hettich Rotors
  • For Use with MIKRO 220/220 R and ROTINA 380/380 R Centrifuges
  • ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards
  • MIKRO 220 and 220 R are powerful, compact benchtop centrifuges for processing microlitre tubes of 0.2 to 2.0mL
Specifications
Specifications
Rotor Adapter
Hettich Drum