Learn More
Hettich™ Adapter for Hettich Drum Rotors no. 1161 and 1711
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1379
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 10226533
  • 1255.00 NOK / Each
Estimated Shipment: 04-06-2024
Description
  • Rotor Carrier for Hettich Rotors
  • For Use with MIKRO 220/220 R and ROTINA 380/380 R Centrifuges
  • ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards
  • MIKRO 220 and 220 R are powerful, compact benchtop centrifuges for processing microlitre tubes of 0.2 to 2.0mL
Specifications
Specifications
Rotor Adapter
32 x 0.2 to 0.4 mL Tubes
43 mm
32
Conical
6 mm
Hettich Drum