Learn More
Hettich™ Adapter for Carrier no. 4285-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 4283-B
Additional Details : Weight : 0.10000kg
Description
- Adapter for Carrier no. 4285-A
- For Deep Well Plates
- Compatible with Swing-out Rotor no. 4282
Specifications
Adapter | |
12 Racks | |
12 Racks per Rotor |
Olympus Racks | |
Carrier no. 4285-A |