Learn More
Hettich™ Adapter for Carrier no. 4285-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
3600.00 NOK - 4790.00 NOK
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
11455028
|
Hettich™
4281 |
16 Plates |
4790.00 NOK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11465028
|
Hettich™
4283-B |
12 Racks |
3600.00 NOK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Adapter for Carrier no. 4285-A
- For Deep Well Plates
- Compatible with Swing-out Rotor no. 4282
Specifications
Adapter | |
Carrier no. 4285-A |