Learn More
Hettich™ Adapter for Carrier no. 4285-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4281
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11455028
  • 4790.00 NOK / Each
Estimated Shipment: 27-06-2024
to see stock.
Description
Description
  • Adapter for Carrier no. 4285-A
  • For Deep Well Plates
  • Compatible with Swing-out Rotor no. 4282
Specifications
Specifications
Adapter
16 Plates
4 to 16 Plates per Rotor
Deep-well Plates
Carrier no. 4285-A