Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Product Code. 13276579 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
for Rotor F-35-6-30, large rotor bore
for Rotor F-35-6-30, small rotor bore
Max. Tube Length:
65 to 89 mm
90 to 110 mm
Unit Size:
Pack of 2
Product Code. For Use With (Equipment) Max. Tube Length unitSize
13276579 for Rotor F-35-6-30, small rotor bore 90 to 110 mm Pack of 2
13216589 for Rotor F-35-6-30, large rotor bore 65 to 89 mm Pack of 2
13286579 for Rotor F-35-6-30, small rotor bore 65 to 89 mm Pack of 2
13256868 for Rotor F-35-6-30, large rotor bore 90 to 110 mm Pack of 2
4 options
Product Code. 13276579
Brand: Eppendorf™ 5427741003
Designed for use with Eppendorf Rotors and Centrifuges
  • For Rotor F-35-6-30
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Max. Tube Length 90 to 110 mm
For Use With (Equipment) for Rotor F-35-6-30, small rotor bore
No. of Places 12