Learn More
Thermo Scientific™ ActivX™ Desthiobiotin-FP Serine Hydrolase Probe
Product Code. 11844624 Shop All Thermo Scientific Products
Click to view available options
Quantity:
4.6 μg
Unit Size:
Each
Product Code. 11844624
Brand: Thermo Scientific™ 88317
ActivX™ probes have been designed to selectively label and enrich serine hydrolases through tagged fluorophosphonate to assess enzyme activity. ActivX Serine Hydrolase Probes covalently label serine hydrolase using fluorophosphonate conjugated with either desthiobiotin or azide.
ActivX™ FP Probes feature a reactive fluorophosphonate group that specifically and covalently labels the active-site serine of enzymatically active serine hydrolases. These probes are available with a desthiobiotin (biotin analog) tag for selective enrichment or a reactive- azido group (Staudinger reagents) that facilitate multiplex labeling when used with phosphine- or alkyne-derivatized tags.
ActivX active-site enrichment probes are advantageous for determining active enzyme levels using analog-tagged probes to allow profiling of both inactive and active enzymes. The ActivX probes selectively enrich only those enzymes that are functionally active and biologically relevant at the time of labeling. With selective enrichment, one can identify and profile target enzyme classes across samples or can assess the specificity and affinity of enzyme inhibitors.
Applications of ActivX FP Probes:
• Determine serine hydrolase enzyme activity in cells and lysates
• Mapping the active-site serine of functionally diverse serine hydrolase family members (e.g. proteases, lipases, esterases)
• Screen for small molecule binding affinities and active-site inhibition
• Profile serine hydrolases using fluorescent, Western blot or mass spectrometry workflows
The serine hydrolase superfamily is one of the largest, most diverse enzyme families in eukaryotic proteomes. Serine hydrolases are generally grouped into two large families: serine proteases (e.g., trypsin, elastase and thrombin) and metabolic serine hydrolases. Metabolic serine hydrolases are divided into multiple enzyme subclasses (e.g., esterases, lipases, amidases and peptidases) based on structure, catalytic mechanism and substrate preference. These probes can be used to assess activity or screen small molecule inhibitors against enzymes derived from cell lysates, subcellular fractions, tissues and recombinant proteins.
Assessment of active-site labeling with desthiobioin probes can be accomplished by either Western blot or mass spectrometry (MS). For the Western blot workflow, desthiobiotin-labeled proteins are enriched for SDS-PAGE analysis and subsequent detection with specific antibodies. For the MS workflow, desthiobiotin-labeled proteins are reduced, alkylated and enzymatically digested to peptides. Only the desthiobiotin-labeled, active-site peptides are enriched for analysis by LC-MS/MS. Both workflows can be used for determining inhibitor target binding, but only the MS workflow can identify global inhibitor targets and off-targets. The Azido-FP probe can be used in combination with a phosphine- or alkyne-derivatized tag for either detection or enrichment of serine hydrolases using Western blot or mass spectrometry.
These products are subject to a limited use label license.
TRUSTED_SUSTAINABILITY
Specifications
Description ActivX™ Desthiobiotin-FP Serine Hydrolase Probe
Formulation Desthiobiotin-fluorophosphonate, MW 464
Content And Storage Store at -80°C away from light. Stock solutions in DMSO can be stored at -20°C for 6 months.
Assay Protein Quantitation Assay
Detection Method Chemiluminescence, Mass Spectrometry, Western Blot
Substrate Type Serine Hydrolase Substrate
Form Lyophilized
Product Line ActivX
Quantity 4.6 μg
For Use With (Application) Pull-Down
Target Serine Hydrolase
Type Fluorophosphonate (FP) probe
Label or Dye Biotin and Other Haptens
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • ActivX Desthiobiotin-FP Serine Hydrolase Probe
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : N-[10-(ethoxy(fluoro)phosphoryl)decyl]-6-[(4R,5S)-5-methyl-2-oxo-imidazolidin-4-yl]hexanamide