Learn More
Honeywell Burdick & Jackson™ Activator Reagent, 0.5M ETT in Acetonitrile
Product Code. p-7161164 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
4 x 2.5 L
Packaging Type:
GLASS BOTTLE, GL45
Unit Size:
2.5 L
Product Code. 17195451
Brand: Honeywell Burdick & Jackson™ BR7252.5GL
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
BioSyn™, for DNA/RNA synthesis
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
CAS 75-05-8
Density 0.809 g/cm3
Packaging Type GLASS BOTTLE, GL45
Quantity 4 x 2.5 L