Learn More
Fisherbrand™ Acrylic Racks for Autosampler Vials
Product Code. 11708256 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W x H):
160 x 160 x 30mm
173 x 91 x 20mm
173 x 95 x 20mm
175.8 x 115.5 x 20mm
Positions:
25
40
50
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Positions Unit Size
11708256 173 x 91 x 20mm 50 Each
11767597 173 x 95 x 20mm 50 Each
11728256 175.8 x 115.5 x 20mm 40 Each
11738256 160 x 160 x 30mm 25 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11708256
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
Easy handling and transportation of sample vials
  • Vial-Rack
  • Acrylic
  • Transparent acrylic material makes filling visible
  • Stable standing position
TRUSTED_SUSTAINABILITY
Specifications
Closure Included No
Autoclavable Not Autoclavable
Color Transparent
Material Acrylic
Positions 50
For Use With (Equipment) ND8 crimp vials
No. per Pack 1
Dimensions (L x W x H) 173 x 91 x 20mm
Product Type Vial Rack