Learn More
Cytiva Acrylic Plastic Thermostat Jacket
Product Code. 10743802 Shop All Cytiva Products
Click to view available options
Diameter (Metric) Outer:
200 mm
300 mm
400 mm
600 mm
700 mm
1000 mm
Diameter (Metric) Inner:
16 mm
26 mm
50 mm
Height (Metric):
200 mm
300 mm
400 mm
600 mm
700 mm
1000 mm
Unit Size:
Each
Product Code. 10743802
Brand: Cytiva 18879101
Only null left
Specifications
For Use With (Equipment) Thermostat
Material Acrylic Plastic
Type Thermostat Jacket
Max. Temperature (Metric) 60°C
Height (Metric) 1000 mm
Diameter (Metric) Inner 26 mm
Diameter (Metric) Outer 1000 mm