missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACF1 Mouse anti-Human, Unconjugated, Clone: 1C4F8, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | ACF1 |
|---|---|
| Clone | 1C4F8 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17501456
|
Proteintech
67947-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17591446
|
Proteintech
67947-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ACF1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ACF1. This antibody recognizes human antigen. The ACF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ACF1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG2b |
| 1C4F8 | |
| Monoclonal | |
| Mouse | |
| 11177 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title