missing translation for 'onlineSavingsMsg'
Learn More

ACF1 Mouse anti-Human, Unconjugated, Clone: 1C4F8, Proteintech

Product Code. 17501456 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17501456 20 μL 20µL
17591446 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17501456

Brand: Proteintech 679471IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The ACF1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ACF1. This antibody recognizes human antigen. The ACF1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen ACF1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C4F8
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 11177
Target Species Human
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.