Learn More
ACF1 Mouse anti-Human, Unconjugated, Clone: 1C4F8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67947-1-IG-150UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17591446
  • 3828.53 NOK / 150µL
Estimated Shipment: 02-10-2023
Description
The ACF1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ACF1. This antibody recognizes human antigen. The ACF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ACF1
Monoclonal
Unconjugated
150 μL
11177
IgG2b
Western Blot, ELISA
1C4F8