missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Acetylcholinesterase/ACHE Assay Kit (Fluorometric)

Product Code. 18643310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18643310 1 Kit 1 set
1 options
This item is not returnable. View return policy

Product Code. 18643310

Brand: Novus Biologicals™ KA4131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assay Kit (Fluorometric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.1 mU/mL to 100 mU/mL
Assay Sensitivity 0.01 mU
Product Type Assay Kit (Fluorometric)
Sample Type Biological Fluids
Specificity Acetylcholinesterase Assay Kit (Green Fluorescence) is a one-step fluorometric assay for the detection of AChE activity. Excitation (Max): 490 nm, Emission (Max): 520 nm.
For Use With (Application) Functional
Gene ID (Entrez) 43
Gene Symbol ACHE
Quantity 1 Kit
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Storage Requirements Store at -20°C.
Synonym acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.