missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Acetylcholinesterase/ACHE Assay Kit (Fluorometric)

Product Code. 18613320 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18613320 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613320 Supplier Novus Biologicals™ Supplier No. KA4132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assay Kit (Fluorometric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.1 to 100 mU/mL
Assay Sensitivity 0.1 mU/mL
Product Type Assay Kit (Fluorometric)
Sample Type Biological Fluids
Specificity The kit is an optimized 'mix and read' assay that provides a simple one- step fluorimetric assay to detect as little as 0.01 mU AChE in a 100 uL assay volume (0.1 mU/mL). Its signal can be easily read with a fluorescence microplate reader at Ex/Em = ∽540/590 nm or an absorbance microplate reader at ∽575 nm.
For Use With (Application) Functional
Gene ID (Entrez) 43
Gene Symbol ACHE
Quantity 1 Kit
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Storage Requirements Store at -20°C.
Synonym acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.