Learn More
Fisherbrand™ accuSolo Clinical Pack
Product Code. 15439719 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
AFS-3/3D
Unit Size:
Each
Product Code. 15439719
Only null left
Add to basket
Add to basket
Uses ion-exchange technology to remove ionic contaminants below trace level. Fisher Scientific™ AccuSolo Clinical Pack provides the water quality required for clinical applications.
  • Removes ionic contaminants
  • Matches water quality required for laboratory applications
Specifications
For Use With (Equipment) AFS-3/3D
Type Filter, Ion-exchange