Learn More
Fisherbrand™ accuQuatro U Pack Cartridge
Product Code. 15419719 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Cartridge Kit with 0.2μm final filter, DI Feed
Cartridge Kit with 0.2μm final filter, RO feed
Cartridge, DI Feed
Cartridge, Low TOC
Cartridge, RO Feed
Unit Size:
Each
Product Code. Type unitSize
15419719 Cartridge, Low TOC Each
15479709 Cartridge, RO Feed Each
15489709 Cartridge Kit with 0.2μm final filter, RO feed Each
15499709 Cartridge, DI Feed Each
15409719 Cartridge Kit with 0.2μm final filter, DI Feed Each
5 options
Product Code. 15419719
Only null left
Add to basket
Add to basket
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
For Use With (Equipment) Accu100
Type Cartridge, Low TOC