Learn More
Fisherbrand™ accuQuatro U Pack Cartridge
Product Code. 15409719 Shop All Fisherbrand Products
Click to view available options
Type:
Cartridge Kit with 0.2μm final filter, DI Feed
Cartridge Kit with 0.2μm final filter, RO feed
Cartridge, DI Feed
Cartridge, Low TOC
Cartridge, RO Feed
Unit Size:
Each
Product Code. 15409719
Only null left
Add to basket
Add to basket
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
For Use With (Equipment) Accu100
Type Cartridge Kit with 0.2μm final filter, DI Feed