Learn More
Fisherbrand™ AccuDuo Cartridge
Product Code. 15499729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
H Pack
P Pack
U Pack
U Pack (low TOC)
U Pack w/ 0.2μm PES Capsule Filter
For Use With (Equipment):
Accu20
Accu20, Accu500
Accu500
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
15499729 U Pack w/ 0.2μm PES Capsule Filter Accu20, Accu500 Each
15459719 H Pack Accu500 Each
15449719 P Pack Accu20 Each
15489729 U Pack Accu20, Accu500 Each
15409739 U Pack (low TOC) Accu20, Accu500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15499729
  • ONLINE EXCLUSIVE PRICE
    4803.15 NOK
    3656.33 NOK / Pack of 1
    Save 1146.82 NOK
    23% Off
View all promotions
Estimated Shipment: 09-03-2027
Add to basket
Add to basket
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
  • Includes two sets of dual-column purification cartridges
  • Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
For Use With (Equipment) Accu20, Accu500
Type U Pack w/ 0.2μm PES Capsule Filter
For Use With (Application) Ultrapure water for critical laboratory applications