Learn More
Miele Professional™ Closed Plinth UG 30-60/60-85
Product Code. 15447724 Shop All Miele Products
Change view
Click to view available options
:
plinth; Dimensions (D x W): 60 x 60mm
Product Code.
15447724 plinth; Dimensions (D x W): 60 x 60mm
1 options
Product Code. 15447724
Brand: Miele Professional™ 12650010_OLD
Only null left
Add to basket
Add to basket
Closed plinth UG 30-60/60-85 to raise the machine, 60 cm deep, 60 cm wide
  • Stainless-steel plinth UG 30-60/60-85
  • For use on PG 8583 and PG 8593 Miele glass laboratory washers
  • Bolts on to machine
  • H 300mm, W 600mm, D 600mm